Learn More
Abcam A23187 (Calcimycin), Ca2+ ionophore, 50MG
Catalog No. 502584633
Encompass_Preferred
MW 523.6 Da, Purity >98%. A mobile ion-carrier that forms stable complexes with divalent cations. Useful for increasing intracellular Ca2+ levels. Also acts as an uncoupler of oxidative phosphorylation and inhibitor of mitochondrial ATPase activity. An antibiotic with weak activity against Gram-positive bacteria.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4633 Supplier Abcam Supplier No. AB12028750MG
MW 523.6 Da, Purity >98%. A mobile ion-carrier that forms stable complexes with divalent cations. Useful for increasing intracellular Ca2+ levels. Also acts as an uncoupler of oxidative phosphorylation and inhibitor of mitochondrial ATPase activity. An antibiotic with weak activity against Gram-positive bacteria.
The product is subject to the following: Abcam Restricted Use Statement