Learn More
Alkali Scientific A23187 [Calcium Ionophore][Calcimycin], 10 Milligrams
SDP
Catalog No. 5000661529
Encompass_Preferred
A23187, also known as Calcimycin, is a calcium ionophore used in biochemical and cellular research to facilitate the transport of calcium ions across biological membranes. This compound increases intracellular calcium levels by forming complexes with divalent cations and enabling their movement through lipid membranes. It is widely used in studies involving signal transduction, cellular activation, and apoptosis, where calcium plays a key regulatory role. A23187 is commonly applied in immunology research to stimulate cells such as lymphocytes and mast cells. It is also used in pharmacological studies investigating calcium-dependent processes, enzyme activation, and membrane transport mechanisms in various biological systems.
Catalog No. 50-006-61529 Supplier Alkali Scientific Supplier No. ACA003CP
A23187, also known as Calcimycin, is a calcium ionophore used in biochemical and cellular research to facilitate the transport of calcium ions across biological membranes. This compound increases intracellular calcium levels by forming complexes with divalent cations and enabling their movement through lipid membranes. It is widely used in studies involving signal transduction, cellular activation, and apoptosis, where calcium plays a key regulatory role. A23187 is commonly applied in immunology research to stimulate cells such as lymphocytes and mast cells. It is also used in pharmacological studies investigating calcium-dependent processes, enzyme activation, and membrane transport mechanisms in various biological systems.