Learn More
Apexbio Technology LLC A23187, free acid 52665-69-7 10mg
SDP
Catalog No. 502240234
Encompass_Preferred
A23187 free acid (CAS 52665-69-7) is a calcium ionophore facilitating Ca transport across biological membranes It is designed to increase intracellular Ca concentration thereby modulating calcium-dependent cellular processes A23187 free acid exerts its biological activity by promoting transmembrane Ca influx In cell-based studies A23187 induces mitochondrial permeability transition and apoptosis in HL-60 cells and stimulates phosphoinositide hydrolysis and inositol release in cultured rat Kupffer cells In rat C6 glioma cells co-treatment with 100 nM A23187 enhances zinc-induced apoptosis with an IC of approximately 1 9 nM correlating with increased intracellular Zn influx Based on these pharmacological properties A23187 free acid holds research potential in studies of apoptosis and calcium or zinc homeostasis
This item is not returnable. View return policy
Catalog No. 50-224-0234 Supplier Apexbio Technology LLC Supplier No. B664610
A23187 free acid (CAS 52665-69-7) is a calcium ionophore facilitating Ca transport across biological membranes It is designed to increase intracellular Ca concentration thereby modulating calcium-dependent cellular processes A23187 free acid exerts its biological activity by promoting transmembrane Ca influx In cell-based studies A23187 induces mitochondrial permeability transition and apoptosis in HL-60 cells and stimulates phosphoinositide hydrolysis and inositol release in cultured rat Kupffer cells In rat C6 glioma cells co-treatment with 100 nM A23187 enhances zinc-induced apoptosis with an IC of approximately 1 9 nM correlating with increased intracellular Zn influx Based on these pharmacological properties A23187 free acid holds research potential in studies of apoptosis and calcium or zinc homeostasis