Learn More
MilliporeSigma A23187 FREE ACID S. CHARTREU
Catalog No. 502303243
Encompass
A23187, Free Acid, Streptomyces chartreusensis; > /=98% by TLC; White solid; EMD; Calbiochem; 5mg; Calcimycin; F.W. 523.6; C9H37N3O6; M.P. 189 deg.C (372.2 deg.F); CAS: 52665-69-7
Catalog No. 50-230-3243 Supplier MilliporeSigma Supplier No. 1001055MG
A23187, Free Acid, Streptomyces chartreusensis; > /=98% by TLC; White solid; EMD; Calbiochem; 5mg; Calcimycin; F.W. 523.6; C9H37N3O6; M.P. 189 deg.C (372.2 deg.F); CAS: 52665-69-7