Learn More
Cayman Chemical A23187calcIM magnsIM sal 10mg
SDP
Catalog No. 502987282
Encompass_Preferred
A divalent cation ionophore, commonly used to facilitate the movement of Ca2+ into cells, triggering the activation of intracellular calcium-dependent pathways; also used to produce apoptosis through calcium overload, as occurs during hypoxic or oxidative stress
Catalog No. 50-298-7282 Supplier Cayman Chemical Supplier No. 2203010 MG
A divalent cation ionophore, commonly used to facilitate the movement of Ca2+ into cells, triggering the activation of intracellular calcium-dependent pathways; also used to produce apoptosis through calcium overload, as occurs during hypoxic or oxidative stress