Learn More
Alkali Scientific A2501 EZ Pack Agarose Tablets
SDP
Catalog No. 503026438
Encompass_Preferred
Researchers trust Alkali Scientifics Agarose LE (A2501) to meet their electrophoreticneeds for a variety of applications Now the same agarose is availablein a convenient no mess tablet Agarose LE is refined in an advanced process that excludes the use oforganic solvents The result is a cleaner end product with significantlyreduced environmental impact The agarose can be used for analyses ofnucleic acids from 150 bp to 25kbp protein electrophoresis and variousblotting protocols The low EEO of the agarose promotes increased electrophoretic mobilityyielding improved resolution and shorter run times This also allowsmacromolecules and larger particles (subcellular fragments viruses etc )to migrate more freely through the gel matrix The consistently low EEOreduces band distortion (caused by counter flow) that can result fromthe presence of excessive sulfate-rich negative ions The quick dissolving EZ Pack tablet contains 0 5g (500mg) ofAgarose LE eliminating the hands-
Catalog No. 50-302-6438 Supplier Alkali Scientific Supplier No. AG2501
Researchers trust Alkali Scientifics Agarose LE (A2501) to meet their electrophoreticneeds for a variety of applications Now the same agarose is availablein a convenient no mess tablet Agarose LE is refined in an advanced process that excludes the use oforganic solvents The result is a cleaner end product with significantlyreduced environmental impact The agarose can be used for analyses ofnucleic acids from 150 bp to 25kbp protein electrophoresis and variousblotting protocols The low EEO of the agarose promotes increased electrophoretic mobilityyielding improved resolution and shorter run times This also allowsmacromolecules and larger particles (subcellular fragments viruses etc )to migrate more freely through the gel matrix The consistently low EEOreduces band distortion (caused by counter flow) that can result fromthe presence of excessive sulfate-rich negative ions The quick dissolving EZ Pack tablet contains 0 5g (500mg) ofAgarose LE eliminating the hands-