Learn More
Cayman Chemical A2764 (hydrochloride), Purity: ≥98%, CAS: 861038-72-4, 10 mg
SDP
Catalog No. 502915495
Encompass_Preferred
An inhibitor of K2P18.1/TRESK (IC50 = 11.8 µM in ionomycin-stimulated Xenopus oocytes expressing the mouse receptor); selective for K2P18.1/TRESK over K2P2.1/TREK1, K2P10.1/TREK2, K2P4.1/TRAAK, K2P3.1/TASK1, K2P3.2/TASK2, K2P3.3/TASK3, K2P16.1/TALK1, and K2P13.1/THIK1 at 100 µM; inhibits basal potassium currents in isolated mouse dorsal root ganglion neurons at 100 µM
Catalog No. 50-291-5495 Supplier Cayman Chemical Supplier No. 3694310 MG
An inhibitor of K2P18.1/TRESK (IC50 = 11.8 µM in ionomycin-stimulated Xenopus oocytes expressing the mouse receptor); selective for K2P18.1/TRESK over K2P2.1/TREK1, K2P10.1/TREK2, K2P4.1/TRAAK, K2P3.1/TASK1, K2P3.2/TASK2, K2P3.3/TASK3, K2P16.1/TALK1, and K2P13.1/THIK1 at 100 µM; inhibits basal potassium currents in isolated mouse dorsal root ganglion neurons at 100 µM