Learn More
Medchemexpress LLC HY-137563 5mg Medchemexpress, A2793 CAS:88349-90-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1375635MG
Encompass_Preferred
Medchemexpress, HY-137563 5mg A2793 CAS:88349-90-0 A2793 is an efficient dual TWIK-related acid-sensitive K+ channel (TASK)-1/TRESK inhibitor, with an IC50 of 6.8 μM for mTRESK. A2764 is more selective for TRESK, and it only moderately influences TREK-1 and TALK-1.
Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1501