Learn More
Cayman Chemical A2AR AgonIst-1 500ug
SDP
Catalog No. 502938559
Encompass_Preferred
An agonist of the adenosine A2A receptor and an inhibitor of ENT1 (Kis = 4.39 and 3.47 µM, for the human receptor and guinea pig transporter, respectively); decreases adenosine uptake by 80% in PC12 cells at 61 µM and increases brain levels of adenosine in a transgenic mouse model of Huntington’s disease at 20 mg/kg; rescues motor performance and increases survival in a transgenic mouse model of Huntington’s disease at 0.11 mg/kg
Catalog No. 50-293-8559 Supplier Cayman Chemical Supplier No. 28414500 UG
An agonist of the adenosine A2A receptor and an inhibitor of ENT1 (Kis = 4.39 and 3.47 µM, for the human receptor and guinea pig transporter, respectively); decreases adenosine uptake by 80% in PC12 cells at 61 µM and increases brain levels of adenosine in a transgenic mouse model of Huntington’s disease at 20 mg/kg; rescues motor performance and increases survival in a transgenic mouse model of Huntington’s disease at 0.11 mg/kg