Learn More
REPLIGEN CORPORATION Repligen ATF2-A2B-TCAQ A2B Tube Set, Designed for Aseptic Connections, Supports High Cell Density Processing, 8.78
SDP
Catalog No. NC3364305
Encompass
The REPLIGEN CORPORATION ATF2-A2B-TCAQ is a durable A2B tube set designed for aseptic connection in XCell ATF 2 bioprocessing systems. It facilitates the sterile transfer of cell culture, supporting high cell densities and continuous processing. Made from AdvantaPure AdvantaSil High Pressure Silicone, it ensures efficient fluid management while maintaining sterility. Its unique design and materials differentiate it from standard tubing kits, making it ideal for demanding cell culture environments.
Catalog No. NC3364305 Supplier REPLIGEN CORPORATION Supplier No. ATF2A2BTCAQ
The REPLIGEN CORPORATION ATF2-A2B-TCAQ is a durable A2B tube set designed for aseptic connection in XCell ATF 2 bioprocessing systems. It facilitates the sterile transfer of cell culture, supporting high cell densities and continuous processing. Made from AdvantaPure AdvantaSil High Pressure Silicone, it ensures efficient fluid management while maintaining sterility. Its unique design and materials differentiate it from standard tubing kits, making it ideal for demanding cell culture environments.