Learn More
AdipoGen A2B2-Ionophore
SDP
Supplier:  AdipoGen CDXA0209M025
Encompass_Preferred
Chemical. CAS 1253421-84-9. Formula C44H36N4O4. MW 684.79. Synthetic. Analytical/Chromatography reagent. Ion sensor compound. Ionophore potentiometric for ISE.
Catalog No. 50-256-1796