Learn More
Aviva Systems Biology Corp A2BP1 Antibody - middle region (ARP58203_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58203_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against A2BP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human A2BP1. Alternate names: FOX1, HRNBP1, 2BP1, A2BP1, FOX-1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Zebrafish
Catalog No. 50-014-53762