Learn More
Aviva Systems Biology Corp A2BP1 Antibody - N-terminal region (ARP58202_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58202_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against A2BP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human A2BP1. Alternate names: FOX1, HRNBP1, 2BP1, A2BP1, FOX-1ApplicationWB. Species reactivity: Cow, Human, Mouse
Catalog No. 50-014-53761