Learn More
Agilent Technologies Glyko A2F (G2FS2), 10µg
Catalog No. NC9698597
Encompass
Glyko Di-sialylated-, galactosylated biantennary complex N-glycan, core-substituted with fucose (A2F). Qualitative standard for glycan identification. Alternative glycan names: G2FS2, FA2G2S2, F(6)A2G(4)2S(6)2
Catalog No. NC9698597 Supplier Agilent Technologies Supplier No. GKC224301
Glyko Di-sialylated-, galactosylated biantennary complex N-glycan, core-substituted with fucose (A2F). Qualitative standard for glycan identification. Alternative glycan names: G2FS2, FA2G2S2, F(6)A2G(4)2S(6)2