Learn More
Aviva Systems Biology Corp A2M Antibody - N-terminal region (ARP59224_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59224_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against A2M. It was validated on Western Blot. . Alternate names: CPAMD5, DKFZp779B086, FWP007, S863-7, A2MDApplicationWB, IHC. Species reactivity: Human
Catalog No. 50-286-621