Learn More
Medchemexpress LLC HY-136465 5mg Medchemexpress, A2ti-1 CAS:570390-00-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1364655MG
Encompass_Preferred
Medchemexpress, HY-136465 5mg A2ti-1 CAS:570390-00-0 A2ti-1 is a selective and high-affinity annexin A2/S100A10 heterotetramer (A2t) inhibitor with an IC50 of 24 μM. A2ti-1 specifically disrupts the protein-protein interaction (PPI) between A2 and S100A10. A2ti-1 prevents human papillomavirus type 16 (HPV16) infection. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1442