Learn More
Medchemexpress LLC HY-136466 5mg Medchemexpress, A2ti-2 CAS:482646-13-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1364665MG
Encompass_Preferred
Medchemexpress, HY-136466 5mg A2ti-2 CAS:482646-13-9 A2ti-2 is a selective and low-affinity annexin A2/S100A10 heterotetramer (A2t) inhibitor with an IC50 of 230 μM. A2ti-2 specifically disrupts the protein-protein interaction (PPI) between A2 and S100A10. A2ti-2 prevents human papillomavirus type 16 (HPV16) infection. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1443