Learn More
MSC PIP Cut & Abrasion-Resistant Gloves, Cut A3, Abrasion 4, 2X-Large
Catalog No. 5000507785
Encompass
These cut and abrasion-resistant gloves are designed for demanding applications. Made with Kevlar and featuring a leather palm coating, they offer protection with a gray on yellow color scheme.
  • Kevlar primary material
  • Leather coating on palm
  • Knit style
  • A3 cut resistance level
  • 4 abrasion resistance level
  • 0 puncture resistance level
  • 2X-Large size
  • 7 gauge
  • Knit cuff style
Catalog No. 50-005-07785 Supplier MSC Supplier No. 89711329
These cut and abrasion-resistant gloves are designed for demanding applications. Made with Kevlar and featuring a leather palm coating, they offer protection with a gray on yellow color scheme.
  • Kevlar primary material
  • Leather coating on palm
  • Knit style
  • A3 cut resistance level
  • 4 abrasion resistance level
  • 0 puncture resistance level
  • 2X-Large size
  • 7 gauge
  • Knit cuff style