Learn More
AdipoGen Melanin Production Inhibitor A3B5 (1 mg)
SDP
Supplier:  AdipoGen AGCR13605M001
Encompass_Preferred
Chemical. CAS: 346721-60-6. Formula: C16H17NO2. MW: 255.3. Non-cytotoxic melanogenesis inhibitor. Potential depigmentation agent. Tyrosinase-related protein 2 (TRP-2) expression inhibitor, via proteasomal degradation. Inhibits the proliferation of melanocytes and melanoma cells through cell cycle arrest in G1. Moderate skin tumor growth suppressor in vivo.
Catalog No. 50-205-3974