Learn More
MSC Honeywell Perfect Fit Gloves, Cut A4, Puncture 3, Abrasion 6, 13 Gauge, Medium
Catalog No. 5000509145
Encompass
The Perfect Fit gloves are engineered to be lightweight, glass fiber-free, and coated with a specialty engineered Polyurethane (PU). This PU coating is designed with a low level of dimethylformamide (DMF) which protects workers at a higher level, while helping them perform at their best with enhanced comfort, dexterity, grip, and protection.
  • Balance protection with dexterity, cut protection, comfort, and grip
  • Low content level of DMF (<100 ppm)
  • Color coded markings
  • QR code with quick access to technical information
  • Silicone-free
  • Coating breathability
  • Glass-fiber free for comfort
  • Lightweight design
  • High grip performance
  • Washable (up to 5 cycles)
Catalog No. 50-005-09145 Supplier MSC Supplier No. 97384200
The Perfect Fit gloves are engineered to be lightweight, glass fiber-free, and coated with a specialty engineered Polyurethane (PU). This PU coating is designed with a low level of dimethylformamide (DMF) which protects workers at a higher level, while helping them perform at their best with enhanced comfort, dexterity, grip, and protection.
  • Balance protection with dexterity, cut protection, comfort, and grip
  • Low content level of DMF (<100 ppm)
  • Color coded markings
  • QR code with quick access to technical information
  • Silicone-free
  • Coating breathability
  • Glass-fiber free for comfort
  • Lightweight design
  • High grip performance
  • Washable (up to 5 cycles)