Learn More
MSC HYFLEX ARAMID C/R GLV
Supplier:  MSC 10174399
Encompass
Cut & Puncture Resistant Gloves; Coating Material: Nitrile ; Coating Coverage: 3/4 Dip and Palm ; Glove Type: Cut-Resistant ; ANSI/ISEA Cut Resistance Level: A4 ; Primary Material: Aramid ; Lining Material: Aramid
Catalog No. 50-273-9154