Learn More
MSC MAXICUT ULTRA NTRL CR GLV
Supplier:  MSC 84424779
Encompass
Cut & Puncture Resistant Gloves; Coating Material: Micro-Foam Nitrile ; Coating Coverage: Palm & Fingers ; Glove Type: Cut-Resistant ; ANSI/ISEA Cut Resistance Level: A4 ; Primary Material: Engineered Yarn ; Lining Material: Engineered Yarn
Catalog No. 50-273-6847