Learn More
MSC PIP TIG Welder's Glove, Kidskin Leather, A4 Cut, 36 cal/cm²
Catalog No. 502834811
Encompass
This TIG welder's glove features lightweight, supple Top Grain Kidskin leather for exceptional dexterity. It includes a Para-Aramid blend lining for A4 cut and heat resistance, and Kevlar stitching for durability. Providing 36 cal/cm² electric arc resistance, it also has an ergonomic keystone thumb for comfort and control, and a split cowhide 5.5 inch leather cuff for forearm protection.
  • Exceptional dexterity with supple kidskin leather.
  • A4 cut and heat resistance from para-aramid lining.
  • Enhanced strength and burn resistance from stitching.
  • Electric arc resistance of 36 cal/cm².
  • Ergonomic keystone thumb for comfort and control.
  • Pulse protector for added safety.
  • 5.5 inch leather cuff for forearm protection and easy wear.
  • Meets Chromium VI free leather requirements.
Catalog No. 50-283-4811 Supplier MSC Supplier No. 90440355
This TIG welder's glove features lightweight, supple Top Grain Kidskin leather for exceptional dexterity. It includes a Para-Aramid blend lining for A4 cut and heat resistance, and Kevlar stitching for durability. Providing 36 cal/cm² electric arc resistance, it also has an ergonomic keystone thumb for comfort and control, and a split cowhide 5.5 inch leather cuff for forearm protection.
  • Exceptional dexterity with supple kidskin leather.
  • A4 cut and heat resistance from para-aramid lining.
  • Enhanced strength and burn resistance from stitching.
  • Electric arc resistance of 36 cal/cm².
  • Ergonomic keystone thumb for comfort and control.
  • Pulse protector for added safety.
  • 5.5 inch leather cuff for forearm protection and easy wear.
  • Meets Chromium VI free leather requirements.