Learn More
Rockland Immunochemicals A431 Whole Cell Lysate EGF Stimulated 500 µg
SDP
Supplier:  Rockland Immunochemicals W09000362
Encompass
Ready-to-use A431 Whole Cell Lysate EGF Stimulated produced by Rockland Immunochemicals are derived from cell lines or tissues using highly refined extraction protocols to ensure exceptionally high quality protein integrity and lot-to-lot reproducibility.  All extracts are tested by SDS-PAGE using 4-20% gradient gels and immunoblot analysis using antibodies to key cell signaling components to confirm the presence of both high molecular weight and low molecular weight proteins.
Catalog No. 50-194-3992