Learn More
Biolegend Alexa Fluor 488 anti-Cyclin B1
Supplier:  Biolegend 647907
Encompass_Preferred
Alexa Fluor 488 anti-Cyclin B1 [V152]; Isotype: Mouse IgG1; Reactivity: Human, Cross-Reactivity: Mouse; Apps: ICFC; Size: 25 tests
Catalog No. 50-208-6959