Learn More
Biolegend Alexa Fluor 488 anti-GFP
Supplier:  Biolegend 338007
Encompass_Preferred
Alexa Fluor 488 anti-GFP [FM264G]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: ICFC, ICC; Size: 25 tests
Catalog No. 50-712-077