Learn More
Biolegend Alexa Fluor 488 anti-H2A.X
Supplier:  Biolegend 600207
Encompass_Preferred
Alexa Fluor 488 anti-H2A.X [W16171A]; Isotype: Rat IgG2a, κ; Reactivity: Human, Mouse; Apps: ICC; Size: 25 ug
Catalog No. 50-208-6616