Learn More
Biolegend Alexa Fluor 488 anti-Histone H3 Phospho (Ser28)
Supplier:  Biolegend 641003
Encompass_Preferred
Alexa Fluor 488 anti-Histone H3 Phospho (Ser28) [HTA28]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-170-939