Learn More
Biolegend Alexa Fluor 488 anti-Histone H3 Phospho (Ser10)
Supplier:  Biolegend 650803
Encompass_Preferred
Alexa Fluor 488 anti-Histone H3 Phospho (Ser10) [11D8]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-171-080