Learn More
Biolegend Alexa Fluor 488 anti-human Ki-67
Supplier:  Biolegend 350532
Encompass_Preferred
Alexa Fluor 488 anti-human Ki-67 [Ki-67]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Bovine; Apps: ICFC, ICC; Size: 100 ug
Catalog No. 50-207-7023