Learn More
Biolegend Alexa Fluor 488 anti-human CD11a/CD18 (LFA-1)
Supplier:  Biolegend 363404
Encompass_Preferred
Alexa Fluor 488 anti-human CD11a/CD18 (LFA-1) [m24]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-103-0821