Learn More
Biolegend Alexa Fluor 488 anti-human CD11c
Supplier:  Biolegend 337236
Encompass_Preferred
Alexa Fluor 488 anti-human CD11c [Bu15]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-6274