Learn More
Biolegend Alexa Fluor 488 anti-human CD154
Supplier:  Biolegend 310815
Encompass_Preferred
Alexa Fluor 488 anti-human CD154 [24-31]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus; Apps: FC, IHC-F; Size: 25 tests
Catalog No. 50-166-568