Learn More
Biolegend Alexa Fluor 488 anti-human CD181 (CXCR1)
Supplier:  Biolegend 320616
Encompass_Preferred
Alexa Fluor 488 anti-human CD181 (CXCR1) [8F1/CXCR1]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-167-029