Learn More
Biolegend Alexa Fluor 488 anti-human CD19
Supplier:  Biolegend 302219
Encompass_Preferred
Alexa Fluor 488 anti-human CD19 [HIB19]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee, Rhesus; Apps: FC, ICC; Size: 100 tests
Catalog No. 50-165-835