Learn More
Biolegend Alexa Fluor 488 anti-human FcεRIα [AER-37 (CRA-1)]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Supplier:  Biolegend 334640
Encompass_Preferred
Alexa Fluor 488 anti-human FcεRIα [AER-37 (CRA-1)]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-237-7281