Learn More
Biolegend Alexa Fluor 488 anti-human IL-17F
Supplier:  Biolegend 516604
Encompass_Preferred
Alexa Fluor 488 anti-human IL-17F [Poly5166]; Isotype: Goat Polyclonal Ig; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-170-204