Learn More
Biolegend Alexa Fluor 488 anti-human IL-2
Supplier:  Biolegend 500314
Encompass_Preferred
Alexa Fluor 488 anti-human IL-2 [MQ1-17H12]; Isotype: Rat IgG2a, κ; Reactivity: Human, Cat (Feline), Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Sooty Mangabey; Apps: ICFC; Size: 100 tests
Catalog No. 50-169-600