Learn More
Biolegend Alexa Fluor 488 anti-human IL-8
Supplier:  Biolegend 511411
Encompass_Preferred
Alexa Fluor 488 anti-human IL-8 [E8N1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-170-093