Learn More
Biolegend Alexa Fluor 488 anti-human TRA-1-81
Supplier:  Biolegend 330709
Encompass_Preferred
Alexa Fluor 488 anti-human TRA-1-81 [TRA-1-81]; Isotype: Mouse IgM, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-5962