Learn More
Biolegend Alexa Fluor 488 anti-human CD185 (CXCR5)
Supplier:  Biolegend 356911
Encompass_Preferred
Alexa Fluor 488 anti-human CD185 (CXCR5) [J252D4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-7606