Learn More
Biolegend Alexa Fluor 488 anti-human CD27 Recombinant
Supplier:  Biolegend 393203
Encompass_Preferred
Alexa Fluor 488 anti-human CD27 Recombinant [QA17A18]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-8971