Learn More
Biolegend Alexa Fluor 488 anti-mouse CD74 (CLIP)
Supplier:  Biolegend 151005
Encompass_Preferred
Alexa Fluor 488 anti-mouse CD74 (CLIP) [In1/CD74]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 25 ug
Catalog No. 50-207-1990