Learn More
Biolegend Alexa Fluor 488 anti-mouse Ki-67
Supplier:  Biolegend 652418
Encompass_Preferred
Alexa Fluor 488 anti-mouse Ki-67 [16A8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 ug
Catalog No. 50-103-0995