Learn More
Biolegend Alexa Fluor 488 anti-mouse CD102
Supplier:  Biolegend 105609
Encompass_Preferred
Alexa Fluor 488 anti-mouse CD102 [3C4 (MIC2/4)]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-163-064