Learn More
Biolegend Alexa Fluor 488 anti-mouse CD147
Supplier:  Biolegend 123718
Encompass_Preferred
Alexa Fluor 488 anti-mouse CD147 [OX-114]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: IHC-F, 3-D IHC; Size: 100 ug
Catalog No. 50-207-0561