Learn More
Biolegend Alexa Fluor 488 anti-mouse CD183 (CXCR3)
Supplier:  Biolegend 126541
Encompass_Preferred
Alexa Fluor 488 anti-mouse CD183 (CXCR3) [CXCR3-173]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-0726