Learn More
Biolegend Alexa Fluor 488 anti-mouse CD19
Supplier:  Biolegend 115521
Encompass_Preferred
Alexa Fluor 488 anti-mouse CD19 [6D5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-604-899