Learn More
Biolegend Alexa Fluor 488 anti-mouse CD28
Supplier:  Biolegend 122017
Encompass_Preferred
Alexa Fluor 488 anti-mouse CD28 [E18]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-005