Learn More
Biolegend Alexa Fluor 488 anti-mouse CD31
Supplier:  Biolegend 102513
Encompass_Preferred
Alexa Fluor 488 anti-mouse CD31 [MEC13.3]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-162-704